2-Fluor-3-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

006422-1G 006422-5G
FLUO006422-1GEA 67.1 EUR
FLUO006422-1G FLUO006422-5G
2-Fluor-3-(trifluormethyl)benzylbromid
2-Fluor-3-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 47…49 °C
MDL: MFCD00061172
CAS-Nummer: 184970-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR