4-Fluor-3-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(Brommethyl)-4-fluor-3-(trifluormethyl)benzol

006428-1G 006428-5G
FLUO006428-1GEA 33 EUR
FLUO006428-1G FLUO006428-5G
4-Fluor-3-(trifluormethyl)benzylbromid
4-Fluor-3-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 22
MDL: MFCD00061177
CAS-Nummer: 184970-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR