2-Methoxy-3-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

CFF659.0.5 CFF659.1 006761-5G 006761-10G 006761-1G 006761-25G
CHMPCFF659.0.5EA 508 EUR
CHMPCFF659.0.5 CHMPCFF659.1 FLUO006761-5G FLUO006761-10G FLUO006761-1G FLUO006761-25G
2-Methoxy-3-(trifluormethyl)pyridin
2-Methoxy-3-(trifluoromethyl)pyridine
Formel: C₇H₆F₃NO
Molecular Weight: 177.13 g/mol
MDL: MFCD00153202
CAS-Nummer: 121643-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR