N-Methyl-2-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

006836-1G 006836-5G
FLUO006836-1GEA 152 EUR
FLUO006836-1G FLUO006836-5G
N-Methyl-2-(trifluormethoxy)anilin
N-Methyl-2-(trifluormethoxy)anilin
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
MDL: MFCD00052334
CAS-Nummer: 175278-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR