6-Nitro-2,3,4-trifluoranilin
Lieferant: FLUOROCHEM
Synonyme:
2-Nitro-4,5,6-trifluoranilin
CFF1249.
006898-1G
006898-5G
CHMPCFF1249.EA
0
EUR
CHMPCFF1249.
FLUO006898-1G
FLUO006898-5G
6-Nitro-2,3,4-trifluoranilin
6-Nitro-2,3,4-trifluoranilin
Formel:
C₆H₃F₃N₂O₂ Molecular Weight: 192,1 g/mol |
MDL:
MFCD00174089 CAS-Nummer: 148416-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...