1-(2-(Trifluormethyl)phenyl)imidazol
Lieferant: FLUOROCHEM
Synonyme:
1-(2-(Trifluoromethyl)phenyl)-1H-imidazole, TRIM
007648-1G
007648-5G
FLUO007648-1GEA
48.8
EUR
FLUO007648-1G
FLUO007648-5G
1-(2-(Trifluormethyl)phenyl)imidazol
1-(2-(Trifluoromethyl)phenyl)imidazole
Formel:
C₁₀H₇F₃N₂ Siedepunkt: 75 °C (0,025 mmHg) Schmelzpunkt: 50…53 °C |
MDL:
MFCD00041206 CAS-Nummer: 25371-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...