Tris(3-fluorphenyl)phosphin

Lieferant: CHEMPUR

FL7751.POR 007751-1G 007751-5G
CHMPFL7751.POREA 0 EUR
CHMPFL7751.POR FLUO007751-1G FLUO007751-5G
Tris(3-fluorphenyl)phosphin
Tris(3-fluorphenyl)phosphin
Formel: C₁₈H₁₂F₃P
Molecular Weight: 316,26 g/mol
MDL: MFCD00015722
CAS-Nummer: 23039-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR