4-Chlor-3-(trifluormethyl)benzylamin

Lieferant: FLUOROCHEM

007809-5G FL7809.10 007809-25G
FLUO007809-5GEA 57.9 EUR
FLUO007809-5G CHMPFL7809.10 FLUO007809-25G
4-Chlor-3-(trifluormethyl)benzylamin
4-Chlor-3-(trifluormethyl)benzylamin
Formel: C₈H₇ClF₃N
Molecular Weight: 209.6 g/mol
Siedepunkt: 100…104 °C (1013 hPa)
Dichte: 1.373
MDL: MFCD00190124
CAS-Nummer: 62039-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR