3-Fluor-4-(trifluormethyl)benzaldehyd
Lieferant: FLUOROCHEM
007907-5G
007907-1G
FLUO007907-5GEA
109
EUR
FLUO007907-5G
FLUO007907-1G
3-Fluor-4-(trifluormethyl)benzaldehyd
3-Fluoro-4-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄F₄O Molecular Weight: 192.11 g/mol |
MDL:
MFCD00236275 CAS-Nummer: 204339-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...