5-Fluor-2-(trifluormethyl)benzaldehyd

Lieferant: FLUOROCHEM

Synonyme: α,α,α,5-Tetrafluoro-o-tolualdehyde, 2-Trifluoromethyl-5-fluorobenzaldehyde

007908-1G 007908-5G 007908-10G
FLUO007908-1GEA 31.1 EUR
FLUO007908-1G FLUO007908-5G FLUO007908-10G
5-Fluor-2-(trifluormethyl)benzaldehyd
5-Fluor-2-(trifluormethyl)benzaldehyd
Formel: C₈H₄F₄O
Molecular Weight: 192.11 g/mol
MDL: MFCD00236276
CAS-Nummer: 90381-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR