4-Fluor-3-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-5-hydroxybenzotrifluoride, 4-Fluoro-3-trifluoromethylphenol

007937-5G 007937-25G
FLUO007937-5GEA 39.8 EUR
FLUO007937-5G FLUO007937-25G
4-Fluor-3-(trifluormethyl)phenol
4-Fluor-3-(trifluormethyl)phenol
Formel: C₇H₄F₄O
Molecular Weight: 180,1 g/mol
Siedepunkt: 84 °C (1013 hPa)
MDL: MFCD00043875
CAS-Nummer: 61721-07-1
EINECS: 432-560-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR