α,α,α-Trifluor-m-xylol
Lieferant: CHEMPUR
Synonyme:
3-(Trifluormethyl)toluol, 3-Methylbenzotrifluorid, 1-Methyl-3-(trifluormethyl)benzol
FL7944.5
FL7944.10
CHMPFL7944.5EA
0
EUR
CHMPFL7944.5
CHMPFL7944.10
α,α,α-Trifluor-m-xylol
α,α,α-Trifluor-m-xylol
Formel:
C₈H₇F₃ Molecular Weight: 160.14 g/mol Siedepunkt: 127 Dichte: 1.148 g/cm³ (20 °C) |
MDL:
MFCD00190105 CAS-Nummer: 401-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...