1-Chlor-3-fluor-4-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-1-fluor-2-iodbenzol, 4-Chlor-2-fluoriodbenzol
008158-10G
008158-25G
FLUO008158-10GEA
0
EUR
FLUO008158-10G
FLUO008158-25G
1-Chlor-3-fluor-4-iodbenzol
1-Chloro-3-fluoro-4-iodobenzene
Formel:
C₆H₃ClFI |
MDL:
MFCD00079729 CAS-Nummer: 6797-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...