2-Fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: (2-Fluorphenyl)metaborsäure, 2-Fluorbenzolboronsäure, Dihydroxy(2-fluorphenyl)boran

008162-25G 008162-5G
FLUO008162-25GEA 73.1 EUR
FLUO008162-25G FLUO008162-5G
2-Fluorphenylboronsäure
2-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
Molecular Weight: 139,92 g/mol
MDL: MFCD00674013
CAS-Nummer: 1993-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR