2,3-Difluor-6-nitrophenol
Lieferant: FLUOROCHEM
Synonyme:
1,2-Difluoro-3-hydroxy-4-nitrobenzene
008167-100G
008167-10G
008167-25G
FLUO008167-100GEA
257
EUR
FLUO008167-100G
FLUO008167-10G
FLUO008167-25G
2,3-Difluor-6-nitrophenol
2,3-Difluor-6-nitrophenol
Formel:
C₆H₃F₂NO₃ Schmelzpunkt: 61 °C |
MDL:
MFCD00042255 CAS-Nummer: 82419-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...