N-Ethyl-2-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethoxy)ethylaminobenzene

008422-1G 008422-5G
FLUO008422-1GEA 72.9 EUR
FLUO008422-1G FLUO008422-5G
N-Ethyl-2-(trifluormethoxy)anilin
N-Ethyl-2-(trifluormethoxy)anilin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
MDL: MFCD00204184
CAS-Nummer: 175278-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR