(±)-2-Butylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: sec-Butylisothiocyanate

008753-1G 008753-5G
FLUO008753-1GEA 51.9 EUR
FLUO008753-1G FLUO008753-5G
(±)-2-Butylisothiocyanat
(±)-2-Butylisothiocyanat
Formel: C₅H₉NS
Molecular Weight: 115,2 g/mol
MDL: MFCD00041108
CAS-Nummer: 4426-79-3
EINECS: 224-609-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR