4-Cyanphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1-Cyan-4-isothiocyanatobenzol, 4-Isothiocyanatobenzoesäurenitril, 4-Isothiocyanatobenzonitril

008781-5G 008781-1G 008781-25G
FLUO008781-5GEA 121 EUR
FLUO008781-5G FLUO008781-1G FLUO008781-25G
4-Cyanphenylisothiocyanat
4-Cyanphenylisothiocyanat
Formel: C₈H₄N₂S
Molecular Weight: 160.2 g/mol
Siedepunkt: 294
Schmelzpunkt: 131…132 °C
MDL: MFCD00041085
CAS-Nummer: 2719-32-6
EINECS: 220-323-2
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR