3-Aminobenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
3-(Hydroxymethyl)anilin, (3-Aminophenyl)methanol
008828-5G
008828-25G
FLUO008828-5GEA
36.9
EUR
FLUO008828-5G
FLUO008828-25G
3-Aminobenzylalkohol
3-Aminobenzylalkohol
Formel:
C₇H₉NO Molecular Weight: 123.15 g/mol Siedepunkt: 272 Schmelzpunkt: 90…93 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007817 CAS-Nummer: 1877-77-6 EINECS: 217-514-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...