3-Methoxybenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Methoxybenzolsulfonsäurechlorid, 3-Methoxybenzolsulfochlorid, 3-Methoxybenzol-1-sulfonylchlorid

008892-1G 008892-5G 008892-25G
FLUO008892-1GEA 140 EUR
FLUO008892-1G FLUO008892-5G FLUO008892-25G
3-Methoxybenzolsulfonylchlorid
3-Methoxybenzolsulfonylchlorid
Formel: C₇H₇ClO₃S
Molecular Weight: 206.65 g/mol
Siedepunkt: 86…98 °C (1 mmHg)
Dichte: 1.46 g/cm³ (20 °C)
MDL: MFCD01318155
CAS-Nummer: 10130-74-2
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR