1-Isopropylpiperazin
Lieferant: FLUOROCHEM
008910-1G
008910-5G
008910-25G
FLUO008910-1GEA
0
EUR
FLUO008910-1G
FLUO008910-5G
FLUO008910-25G
1-Isopropylpiperazin
1-Isopropylpiperazin
Formel:
C₇H₁₆N₂ |
MDL:
MFCD00167971 CAS-Nummer: 4318-42-7 EINECS: 224-345-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...