2-Amino-4-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine
009013-500MG
009013-1G
FLUO009013-500MGEA
0
EUR
FLUO009013-500MG
FLUO009013-1G
2-Amino-4-(trifluormethyl)pyridin
2-Amino-4-(trifluoromethyl)pyridine
Formel:
C₆H₅F₃N₂ Molecular Weight: 162,11 g/mol |
MDL:
MFCD00077455 CAS-Nummer: 106447-97-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...