4-Amino-3-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
009014-1G
FL9014.5
009014-500MG
FLUO009014-1GEA
525
EUR
FLUO009014-1G
CHMPFL9014.5
FLUO009014-500MG
4-Amino-3-(trifluormethyl)pyridin
4-Amino-3-(trifluoromethyl)pyridine
Formel:
C₆H₅F₃N₂ Molecular Weight: 162,11 g/mol |
MDL:
MFCD02183554 CAS-Nummer: 387824-61-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...