5-Amino-2-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluormethyl)pyridin-5-amin, 2-(Trifluormethyl)-5-pyridylamin

009015-1G 009015-5G
FLUO009015-1GEA 30.8 EUR
FLUO009015-1G FLUO009015-5G
5-Amino-2-(trifluormethyl)pyridin
5-Amino-2-(trifluormethyl)pyridin
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
MDL: MFCD01862656
CAS-Nummer: 106877-33-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR