4-Chlor-3-fluorbenzotrifluorid
Lieferant: FLUOROCHEM
009083-5G
009083-10G
FLUO009083-5GEA
311
EUR
FLUO009083-5G
FLUO009083-10G
4-Chlor-3-fluorbenzotrifluorid
4-Chlor-3-fluorbenzotrifluorid
Formel:
C₇H₃ClF₄ Molecular Weight: 198,55 g/mol |
MDL:
MFCD00060646 CAS-Nummer: 32137-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...