3,3'-Difluorbenzophenon

Lieferant: FLUOROCHEM

Synonyme: Bis(3-fluorphenyl)keton

009139-1G 009139-5G
FLUO009139-1GEA 112 EUR
FLUO009139-1G FLUO009139-5G
3,3'-Difluorbenzophenon
3,3'-Difluorbenzophenon
Formel: C₁₃H₈F₂O
MDL: MFCD00000337
CAS-Nummer: 345-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR