1-(2,4-Difluorphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-(2,4-Difluorphenyl)thioharnstoff, 2,4-Difluorphenylthioharnstoff

009148-5G 009148-25G
FLUO009148-5GEA 72.9 EUR
FLUO009148-5G FLUO009148-25G
1-(2,4-Difluorphenyl)-2-thioharnstoff
1-(2,4-Difluorphenyl)-2-thioharnstoff
Formel: (C₆H₃F₂)NC(S)NH₂
Molecular Weight: 188,2 g/mol
Siedepunkt: 272 °C (1013 hPa)
Schmelzpunkt: 165…166 °C
MDL: MFCD00041150
CAS-Nummer: 175277-76-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR