3-(2-Fluorphenyl)propionsäure

Lieferant: FLUOROCHEM

009235-5G 009235-25G 009235-1G
FLUO009235-5GEA 79.1 EUR
FLUO009235-5G FLUO009235-25G FLUO009235-1G
3-(2-Fluorphenyl)propionsäure
3-(2-Fluorphenyl)propionsäure
Formel: C₉H₉FO₂
Molecular Weight: 168.17 g/mol
MDL: MFCD01310820
CAS-Nummer: 1643-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR