3-(3-Fluorphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Fluorphenyl)propansäure

009236-1G 009236-5G
FLUO009236-1GEA 53.9 EUR
FLUO009236-1G FLUO009236-5G
3-(3-Fluorphenyl)propionsäure
3-(3-Fluorphenyl)propionsäure
Formel: C₉H₉FO₂
Molecular Weight: 168.17 g/mol
MDL: MFCD01310821
CAS-Nummer: 458-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR