4-(Trifluormethyl)pyridin

Lieferant: FLUOROCHEM

009275-1G 009275-5G 009275-25G
FLUO009275-1GEA 36.9 EUR
FLUO009275-1G FLUO009275-5G FLUO009275-25G
4-(Trifluormethyl)pyridin
4-(Trifluoromethyl)pyridine
Formel: C₆H₄F₃N
Molecular Weight: 147,1 g/mol
Siedepunkt: 105…106 °C (1013 hPa)
MDL: MFCD00153289
CAS-Nummer: 3796-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR