2-Chlor-6-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

009416-1G 009416-5G 009416-25G
FLUO009416-1GEA 0 EUR
FLUO009416-1G FLUO009416-5G FLUO009416-25G
2-Chlor-6-(trifluormethyl)pyridin
2-Chloro-6-(trifluoromethyl)pyridine
Formel: C₆H₃ClF₃N
Molecular Weight: 181.54 g/mol
MDL: MFCD00042226
CAS-Nummer: 39890-95-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR