2-Amino-6-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

009417-500MG 009417-1G
FLUO009417-500MGEA 0 EUR
FLUO009417-500MG FLUO009417-1G
2-Amino-6-(trifluormethyl)pyridin
2-Amino-6-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
Siedepunkt: 200 °C (1013 hPa)
Schmelzpunkt: 78…80 °C
MDL: MFCD03093868
CAS-Nummer: 34486-24-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR