4-Methyl-2-nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

009540-1G 009540-5G
FLUO009540-1GEA 76.2 EUR
FLUO009540-1G FLUO009540-5G
4-Methyl-2-nitrophenylisothiocyanat
4-Methyl-2-nitrophenylisothiocyanat
Formel: C₈H₆N₂O₂S
Molecular Weight: 194,21 g/mol
Schmelzpunkt: 67 °C
MDL: MFCD00041248
CAS-Nummer: 17614-74-3
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR