2-Isopropyl-4-methylthiazol

Lieferant: FLUOROCHEM

009624-100G 009624-25G
FLUO009624-100GEA 206 EUR
FLUO009624-100G FLUO009624-25G
2-Isopropyl-4-methylthiazol
2-Isopropyl-4-methylthiazol
Formel: C₇H₁₁NS
Molecular Weight: 141,24 g/mol
MDL: MFCD00036561
CAS-Nummer: 15679-13-7
EINECS: 239-758-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR