3-(Difluormethoxy)anilin

Lieferant: FLUOROCHEM
Achtung

CFF1035. 009628-25G 009628-5G
CHMPCFF1035.EA 0 EUR
CHMPCFF1035. FLUO009628-25G FLUO009628-5G
3-(Difluormethoxy)anilin
3-(Difluormethoxy)anilin
Formel: C₇H₇F₂NO
Molecular Weight: 159.14 g/mol
MDL: MFCD00236221
CAS-Nummer: 22236-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR