4-Brom-N-methylanilin

Lieferant: FLUOROCHEM

009647-10G 009647-1G 009647-25G 009647-5G
FLUO009647-10GEA 0 EUR
FLUO009647-10G FLUO009647-1G FLUO009647-25G FLUO009647-5G
4-Brom-N-methylanilin
4-Brom-N-methylanilin
Formel: C₇H₈BrN
Molecular Weight: 186.05 g/mol
MDL: MFCD03030391
CAS-Nummer: 6911-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR