2-Chlor-N-methylanilin
Lieferant: FLUOROCHEM
009648-10G
009648-25G
009648-5G
FLUO009648-10GEA
224
EUR
FLUO009648-10G
FLUO009648-25G
FLUO009648-5G
2-Chlor-N-methylanilin
2-Chlor-N-methylanilin
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 95…96 °C (20 mmHg) Dichte: 1,15 g/cm³ (20 °C) |
MDL:
MFCD00045170 CAS-Nummer: 932-32-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...