3-Chlor-N-methylanilin
Lieferant: FLUOROCHEM
Synonyme:
N-Methyl-3-chloroaniline
009649-10G
009649-25G
009649-5G
FLUO009649-10GEA
174
EUR
FLUO009649-10G
FLUO009649-25G
FLUO009649-5G
3-Chlor-N-methylanilin
3-Chlor-N-methylanilin
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 235 °C (1013 hPa) Dichte: 1,174 g/cm³ (11.5 °C) |
MDL:
MFCD00052014 CAS-Nummer: 7006-52-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...