5-(Chlormethyl)uracil

Lieferant: CHEMPUR

Synonyme: 5-(Chlormethyl)-2,4-dihydroxypyrimidin

FL9757.1 FL9757.5
CHMPFL9757.1EA 0 EUR
CHMPFL9757.1 CHMPFL9757.5
5-(Chlormethyl)uracil
5-(Chlormethyl)uracil
Formel: C₅H₅ClN₂O₂
Molecular Weight: 160.56 g/mol
MDL: MFCD00218445
CAS-Nummer: 3590-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR