1-(4-Methylphenyl)piperazin

Lieferant: FLUOROCHEM

Synonyme: 1-(p-Tolyl)piperazin

009809-5G 009809-25G
FLUO009809-5GEA 0 EUR
FLUO009809-5G FLUO009809-25G
1-(4-Methylphenyl)piperazin
1-(4-Methylphenyl)piperazin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
MDL: MFCD00040737
CAS-Nummer: 39593-08-3
EINECS: 254-534-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR