2-Thenoesäurehydrazid
Lieferant: CHEMPUR
Synonyme:
2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid
FL9911.POR
009911-10G
009911-25G
CHMPFL9911.POREA
0
EUR
CHMPFL9911.POR
FLUO009911-10G
FLUO009911-25G
2-Thenoesäurehydrazid
2-Thenoesäurehydrazid
Formel:
C₅H₆N₂OS Molecular Weight: 142,18 g/mol Schmelzpunkt: 142…143 °C |
MDL:
MFCD00005435 CAS-Nummer: 2361-27-5 EINECS: 219-107-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...