5-Fluor-3-indolylessigsäure

Lieferant: FLUOROCHEM

010188-1G 010188-500MG
FLUO010188-1GEA 527 EUR
FLUO010188-1G FLUO010188-500MG
5-Fluor-3-indolylessigsäure
5-Fluor-3-indolylessigsäure
Formel: C₁₀H₈FNO₂
Molecular Weight: 193.18 g/mol
MDL: MFCD00056921
CAS-Nummer: 443-73-2
EINECS: 207-138-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR