5-Fluor-2-indolcarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Fluoro-1H-indole-2-carboxylic acid

CFF1112. 010193-10G 010193-1G
CHMPCFF1112.EA 0 EUR
CHMPCFF1112. FLUO010193-10G FLUO010193-1G
5-Fluor-2-indolcarbonsäure
5-Fluoroindole-2-carboxylic acid
Formel: C₉H₆FNO₂
Schmelzpunkt: 258…259 °C
MDL: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR