2,6-Bis(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

010225-1G 010225-5G
FLUO010225-1GEA 57.8 EUR
FLUO010225-1G FLUO010225-5G
2,6-Bis(trifluormethyl)pyridin
2,6-Bis(trifluormethyl)pyridin
Formel: C₇H₃F₆N
Molecular Weight: 215.1 g/mol
MDL: MFCD00236675
CAS-Nummer: 455-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR