4-Fluorsalicylsäure
Lieferant: FLUOROCHEM
Synonyme:
4-Fluoro-2-hydroxybenzoic acid
010305-1G
010305-5G
FLUO010305-1GEA
31.1
EUR
FLUO010305-1G
FLUO010305-5G
4-Fluorsalicylsäure
4-Fluorsalicylsäure
Formel:
C₇H₅FO₃ Molecular Weight: 156,11 g/mol |
MDL:
MFCD00153168 CAS-Nummer: 345-29-9 EINECS: 206-459-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...