2,3,5,6-Tetrafluor-p-toluylsäure
Lieferant: FLUOROCHEM
010542-1G
010542-5G
FLUO010542-1GEA
54.8
EUR
FLUO010542-1G
FLUO010542-5G
2,3,5,6-Tetrafluor-p-toluylsäure
2,3,5,6-Tetrafluor-p-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208,11 g/mol |
MDL:
MFCD00012302 CAS-Nummer: 652-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...