6-Fluor-m-toluonitril

Lieferant: FLUOROCHEM

010683-1G 010683-5G
FLUO010683-1GEA 31.1 EUR
FLUO010683-1G FLUO010683-5G
6-Fluor-m-toluonitril
6-Fluor-m-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
MDL: MFCD00134542
CAS-Nummer: 64113-84-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR