2-(3-Fluorphenyl)ethanol

Lieferant: FLUOROCHEM

010684-1G 010684-5G
FLUO010684-1GEA 36.9 EUR
FLUO010684-1G FLUO010684-5G
2-(3-Fluorphenyl)ethanol
2-(3-Fluorphenyl)ethanol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
Siedepunkt: 70…71 °C (2 mmHg)
Dichte: 1.12 g/cm³ (20 °C)
MDL: MFCD00045998
CAS-Nummer: 52059-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR