3-(2-Aminoethyl)pyridin

Lieferant: FLUOROCHEM

010689-1G 010689-5G
FLUO010689-1GEA 66.5 EUR
FLUO010689-1G FLUO010689-5G
3-(2-Aminoethyl)pyridin
3-(2-Aminoethyl)pyridine
Formel: C₇H₁₀N₂
Siedepunkt: 54 °C (0,06 mmHg)
MDL: MFCD00191604
CAS-Nummer: 20173-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR