2-Amino-5-chlor-3-nitropyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 5-Chlor-3-nitropyridin-2-amin, 5-Chlor-3-nitro-2-pyridylamin

010693-5G 010693-25G
FLUO010693-5GEA 0 EUR
FLUO010693-5G FLUO010693-25G
2-Amino-5-chlor-3-nitropyridin
2-Amino-5-chlor-3-nitropyridin
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173,56 g/mol
MDL: MFCD00092011
CAS-Nummer: 5409-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR